Anti-KCNMB1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A55547-100UG A55547-50UG
77824-585EA 521.4 CAD
77824-585 77824-586
Anti-KCNMB1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: KCNMB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR