Anti-ALOX15B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52935-100UL A52935-50UL
77819-874EA 521.4 CAD
77819-874 77819-875
Anti-ALOX15B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:15 to 1:50

Type:
Antigen: ALOX15B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR