Anti-ARMC10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A52268-100UL A52268-50UL
77818-660EA 521.4 CAD
77818-660 77818-661
Anti-ARMC10 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: ARMC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR