Anti-BMP4 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A41314-100UG A41314-50UG
77808-944EA 521.4 CAD
77808-944 77808-945
Anti-BMP4 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: BMP4
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol BMP4
Environmentally Preferable
Antigen name Bone morphogenetic protein 4 protein (303-408AA)
Antigen synonyms zgc:100779|BMP 2B|BMP 4|BMP-2B|BMP-4|BMP2B|BMP2B1|BMP4|BMP4_HUMAN|Bone morphogenetic protein 2B|Bone morphogenetic protein 4|DVR4|MCOPS6|MGC100779|OFC11|zbmp-4|ZYME
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P12644
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR