Anti-GBE1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A58793-100UL A58793-50UL
77830-686EA 521.4 CAD
77830-686 77830-687
Anti-GBE1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200

Type:
Antigen: GBE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR