Anti-PTPRJ Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A59955-100UL A59955-50UL
77832-857EA 521.4 CAD
77832-857 77832-858
Anti-PTPRJ Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: PTPRJ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR