Anti-Reg3g Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A33043-100UG A33043-50UG
77797-011EA 521.4 CAD
77797-011 77797-012
Anti-Reg3g Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: Reg3g
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
New Product
Order Now

SPECIFICATIONS

Antigen symbol Reg3g
Antigen name Recombinant Rat Regenerating islet-derived protein 3-gamma protein (27-174AA)
Environmentally Preferable
Antigen synonyms Reg3g| Pap3| Regenerating islet-derived protein 3-gamma| REG-3-gamma| Pancreatitis-associated protein 3| Regenerating islet-derived protein III-gamma| Reg III-gamma) [Cleaved into: Regenerating islet-derived protein 3-gamma 16.5 kDa form| Regenerating islet-derived protein 3-gamma 15 kDa form]
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Rat
Host Rabbit
Gene ID P42854
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR