Anti-III Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A50060-100UG A50060-50UG
77814-580EA 521.4 CAD
77814-580 77814-581
Anti-III Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type:
Antigen: III
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterobacteria phage M13
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR