Anti-POMGNT1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A65783-100UG A65783-50UG
77833-730EA 521.4 CAD
77833-730 77833-731
Anti-POMGNT1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: POMGNT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR