Anti-NUDT9 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A69295-100UG A69295-50UG
77840-332EA 521.4 CAD
77840-332 77840-333
Anti-NUDT9 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NUDT9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR