Anti-Sialic acid-binding lectin Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A50467-100UG A50467-50UG
77815-349EA 521.4 CAD
77815-349 77815-350
Anti-Sialic acid-binding lectin Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Type:
Antigen: Sialic acid-binding lectin
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rana japonica
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR