Anti-THBS1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A42632-100UG A42632-50UG
77810-832EA 521.4 CAD
77810-832 77810-833
Anti-THBS1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: THBS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR