Anti-SLC35D3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A35015-100UG A35015-50UG
77800-039EA 521.4 CAD
77800-039 77800-040
Anti-SLC35D3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:50 to 1:200

Type:
Antigen: SLC35D3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR