Anti-PDE6H Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A54719-100UG A54719-50UG
77823-024EA 521.4 CAD
77823-024 77823-025
Anti-PDE6H Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: PDE6H
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR