Anti-PEMT Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A30951-100UG A30951-50UG
77793-708EA 521.4 CAD
77793-708 77793-709
Anti-PEMT Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PEMT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR