Anti-glpD Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A50385-100UG A50385-50UG
77815-189EA 521.4 CAD
77815-189 77815-190
Anti-glpD Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: glpD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
New Product
Order Now

SPECIFICATIONS

Antigen symbol glpD
Environmentally Preferable
Antigen name Recombinant Escherichia coli Aerobic glycerol-3-phosphate dehydrogenase protein (1-501AA)
Antigen synonyms glpD|glyD|b3426|JW3389Aerobic glycerol-3-phosphate dehydrogenase|EC 1.1.5.3
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity E. coli
Host Rabbit
Gene ID P13035
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR