Anti-KCNC3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55435-100UG A55435-50UG
77824-370EA 521.4 CAD
77824-370 77824-371
Anti-KCNC3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: KCNC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR