Anti-SLC6A12 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A35073-50UL A35073-100UL
77800-134EA 355.5 CAD
77800-134 77800-133
Anti-SLC6A12 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: SLC6A12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR