Anti-DLL3 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A69708-100UG A69708-50UG
77841-114EA 521.4 CAD
77841-114 77841-115
Anti-DLL3 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: DLL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR