Anti-HNRNPH1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A40434-100UG A40434-50UG
77807-622EA 521.4 CAD
77807-622 77807-623
Anti-HNRNPH1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: HNRNPH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR