Anti-GCY1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50420-100UG A50420-50UG
77815-256EA 521.4 CAD
77815-256 77815-257
Anti-GCY1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: GCY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR