Anti-NS Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51296-100UG A51296-50UG
77816-909EA 521.4 CAD
77816-909 77816-910
Anti-NS Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: NS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR