Anti-IDH3B Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A41193-100UG A41193-50UG
77808-727EA 521.4 CAD
77808-727 77808-728
Anti-IDH3B Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: IDH3B
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR