Anti-egsA Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A52345-100UG A52345-50UG
77818-794EA 521.4 CAD
77818-794 77818-795
Anti-egsA Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: egsA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Geobacillus stearothermophilus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR