Anti-GPAM Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A68888-100UG A68888-50UG
77839-571EA 521.4 CAD
77839-571 77839-572
Anti-GPAM Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:200 to 1:3000, IHC: 1:20 to 1:200

Type:
Antigen: GPAM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR