Anti-L-lactate dehydrogenase Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A56073-100UG A56073-50UG
77825-588EA 521.4 CAD
77825-588 77825-589
Anti-L-lactate dehydrogenase Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: L-lactate dehydrogenase
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR