Anti-CSTA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A56082-100UL A56082-50UL
77825-605EA 521.4 CAD
77825-605 77825-606
Anti-CSTA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000

Type:
Antigen: CSTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR