Anti-RNF26 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A33367-100UG A33367-50UG
77797-540EA 521.4 CAD
77797-540 77797-541
Anti-RNF26 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RNF26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR