Anti-MAGI1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66095-100UG A66095-50UG
77834-324EA 521.4 CAD
77834-324 77834-325
Anti-MAGI1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: MAGI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR