Anti-PSMA2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A32232-100UG A32232-50UG
77795-755EA 521.4 CAD
77795-755 77795-756
Anti-PSMA2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PSMA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR