Anti-PTPN22 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A32449-100UL A32449-50UL
77796-099EA 521.4 CAD
77796-099 77796-100
Anti-PTPN22 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: PTPN22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR