Anti-SLC37A1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35025-100UG A35025-50UG
77800-059EA 521.4 CAD
77800-059 77800-060
Anti-SLC37A1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC37A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR