Anti-P Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50552-100UG A50552-50UG
77815-508EA 521.4 CAD
77815-508 77815-509
Anti-P Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hepatitis B virus genotype A2 subtype adw2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR