Anti-SLC25A18 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A69414-50UG A69414-100UG
77840-558EA 355.5 CAD
77840-558 77840-557
Anti-SLC25A18 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC25A18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR