Anti-PDE8B Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A52756-100UL A52756-50UL
77819-561EA 521.4 CAD
77819-561 77819-562
Anti-PDE8B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: PDE8B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR