Anti-MS4A6E Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A65932-100UG A65932-50UG
77834-013EA 521.4 CAD
77834-013 77834-014
Anti-MS4A6E Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MS4A6E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR