Anti-fabI Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A58560-100UG A58560-50UG
77830-238EA 521.4 CAD
77830-238 77830-239
Anti-fabI Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: fabI
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR