Anti-FAM53B Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A54734-50UG A54734-100UG
77823-052EA 355.5 CAD
77823-052 77823-051
Anti-FAM53B Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FAM53B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR