Anti-PAN2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A56676-100UG A56676-50UG
77826-717EA 521.4 CAD
77826-717 77826-718
Anti-PAN2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: PAN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR