Anti-MIA Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A54300-100UG A54300-50UG
77822-233EA 521.4 CAD
77822-233 77822-234
Anti-MIA Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: MIA
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR