Anti-Histone H2B Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A51262-100UG A51262-50UG
77816-841EA 521.4 CAD
77816-841 77816-842
Anti-Histone H2B Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Histone H2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cairina moschata
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR