Anti-dye Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A59666-100UG A59666-50UG
77832-325EA 521.4 CAD
77832-325 77832-326
Anti-dye Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: dye
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR