Anti-BG2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A50678-100UL A50678-50UL
77815-750EA 521.4 CAD
77815-750 77815-751
Anti-BG2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: BG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR