Anti-GLO1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66865-100UL A66865-50UL
77835-769EA 521.4 CAD
77835-769 77835-770
Anti-GLO1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: GLO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR