Anti-MTBP Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A65940-100UG A65940-50UG
77834-029EA 521.4 CAD
77834-029 77834-030
Anti-MTBP Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500

Type:
Antigen: MTBP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR