Anti-GK3P Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A54442-100UG A54442-50UG
77822-502EA 521.4 CAD
77822-502 77822-503
Anti-GK3P Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GK3P
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR