Anti-CASP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A58968-100UL A58968-50UL
77831-015EA 521.4 CAD
77831-015 77831-016
Anti-CASP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:10000, WB: 1:1000 to 1:4000, IHC: 1:25 to 1:100

Type:
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR