Anti-PDE6C Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A58851-100UL A58851-50UL
77830-795EA 521.4 CAD
77830-795 77830-796
Anti-PDE6C Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000

Type:
Antigen: PDE6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR