Anti-RBP4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A42498-50UG A42498-100UG
77810-590EA 355.5 CAD
77810-590 77810-589
Anti-RBP4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: RBP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol RBP4
Environmentally Preferable
Antigen name Retinol-binding protein 4 protein (19-201AA)
Antigen synonyms OTTHUMP00000020114| OTTHUMP00000020115| OTTHUMP00000020116| Plasma retinol binding protein 4| Plasma retinol-binding protein| Plasma retinol-binding protein(1-176)| PRBP| PRO2222| RBP| RBP4| RDCCAS| RET4_HUMAN| Retinol binding protein 4| Retinol binding protein 4 interstitial| Retinol binding protein 4 plasma
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P02753
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR