Anti-ZNF221 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A38795-100UG A38795-50UG
77805-143EA 521.4 CAD
77805-143 77805-144
Anti-ZNF221 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ZNF221
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR